TISSUE-TROL™ Control Slides Argentaffin
Leverancier: Sigma Aldrich
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
TISSUE-TROL™
TTR013-25EAEA 288 EUR
TTR013-25EA
TISSUE-TROL™ Control Slides Argentaffin
Object- en dekglaasjes Microscopy Control Slides
Histology slides with paraffin embedded section of human intestine containing argentaffin granules. Suitable for use with Fontana-Masson stain kit (Procedure No. HT200).
Leveringsinformatie: Box contains 24 unstained slides plus one reference slide.