Eosin Y (yellowish) 0,5% in aqueous solution for microscopy, Sigma-Aldrich®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Solvent Red 43, Field's Stain B, Eosin 225, 2,4,5,7-Tetrabromofluorescein disodium salt, Disodium 2-(2,4,5,7-tetrabromo-6-oxido-3-oxoxanthen-9-yl)benzoate, Disodium 2',4',5',7'-tetrabromo-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-3',6'-bis(olate), Tetrabromofluorescein disodium salt, 2′,4′,5′,7′-Tetrabromofluorescein disodium salt, Eosin Y disodium salt
1.09844.1000 1.09844.2500
1.09844.1000EA 85.3 EUR
1.09844.1000 1.09844.2500
Eosin Y (yellowish) 0,5% in aqueous solution for microscopy, Sigma-Aldrich®
Eosin Y (yellowish)
Formule: C₂₀H₆Br₄Na₂O₅
MW: 691.86 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00005040
CAS nummer: 17372-87-1
EINECS: 241-409-6
Merck Index: 13,03634
IVDLogo