Turkey red oil sodium salt Microscopy, Sigma-Aldrich®
Leverancier: SIGMA ALDRICH MICROSCOPY
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Sulphated castor oil, Sulphoricinolate sodium salt, Castor-oil sulphated sodium salt
MQ100
16316-1L 16316-30KG
16316-1LEA 103 EUR
16316-1L 16316-30KG
Turkey red oil sodium salt Microscopy, Sigma-Aldrich®
Turkey red oil sodium salt
  • assay 100%
  • impurities 16-20% water (Karl Fischer)
Dichtheid: 1.05 g/cm³ (20 °C)
Vlampunt: 109 °C (gesloten beker)
MDL Nummer: MFCD00132540
CAS nummer: 68187-76-8
EINECS: 232-306-7