CleanMax® Clean Manufactured Coveralls with Attached Hood and Boots
Leverancier: LAKELAND CLEANMAX APPAREL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CTL414CM-2X CTL414CM-3X CTL414CM-4X CTL414CM-5X CTL414CM-LG CTL414CM-MD CTL414CM-XL CTL414CM-SM
114-9570EA 499 EUR
114-9570 114-9571 114-9572 114-9573 114-9574 114-9575 114-9576 CLLLCTL414CM-SM
CleanMax® Clean Manufactured Coveralls with Attached Hood and Boots
Wegwerpkleding
Cleanroom garments are constructed and packaged to offer premium protection for your cleanroom and controlled environments.
  • Attached hood
  • Attached boots
  • Elastic wrist
Cleanroom coveralls with attached hood and boots is ideal for ISO Class 4 to 6 cleanrooms.
Verpakking: These products are offered in a case of 25.