Water Separator
Leverancier: NEUBERT VOLUME GLASSWAERE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
-90-10292914 -90-10292919 03829010292919 03829010292914
NEUB-90-10292914EA 424 EUR
NEUB-90-10292914 NEUB-90-10292919 NEUB03829010292919 NEUB03829010292914
Water Separator
Onderdelen voor destillatie Destillatiesets
Water separator made of Borosilicate glass 3.3.
  • Graduation 10 ml in 1/10, 0-point on the stopcock
  • With 2-way tap