Human Tagged ORF Clone, Androgen Receptor (AR) (NM_000044)
Leverancier: OriGene
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ORIGRC235415EA 1700 EUR
ORIGRC235415
Human Tagged ORF Clone, Androgen Receptor (AR) (NM_000044)
Vectoren, Plasmiden en Bibliotheken
The ORF clone is ion-exchange column purified and shipped in a 2D barcoded Matrix tube containing 10 µg of transfection-ready, dried plasmid DNA (reconstitute with 100 µl of water).
  • Symbol: Androgen receptor
  • Synonyms: AIS; AR8; DHTR; HUMARA; HYSP1; KD; NR3C4; SBMA; SMAX1; TFM
  • E. coli selection: Kanamycin (25 µg/ml)
  • Mammalian cell selection: Neomycin
  • ORF size: 2760 bp
  • UniProt ID: P10275
  • Cytogenetics: Xq12
  • Domains: HOLI, Androgen_recep, zf-C4
  • Molecular weight: 99,2 kDa