Gloves, Nitrile, Powder-Free, Coverfeel Touch
Leverancier: Medicom
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1140-C 1140-D 1140-E 1140-A 1140-B
HOPE1140-CEA 156 EUR
HOPE1140-C HOPE1140-D HOPE1140-E HOPE1140-A HOPE1140-B
Gloves, Nitrile, Powder-Free, Coverfeel Touch
Handschoenen
Kolmi Coverfeel Touch glove Class I - Personal Protective Equipment, nitrile, powder-free and textured fingertips.
  • No accelerator to limit type IV allergies
  • Ultra-comfortable glove and faster donning thanks to its internal polymer coating
  • Great elasticity and good dexterity
  • Non-slip texture for excellent grip and handling of objects
  • Made in France
Recommended in medical, industrial and agri-food environments.
Certificaten: EN ISO 374-1 Type B, ASTM D6978-05, EN ISO 374-5:2016.