Gloves, Nitrile, Powder-Free, Coverfeel Care Long
Leverancier: Medicom
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1144-D 1144-E 1144-F 1144-B 1144-C
HOPE1144-DEA 253 EUR
HOPE1144-D HOPE1144-E HOPE1144-F HOPE1144-B HOPE1144-C
Gloves, Nitrile, Powder-Free, Coverfeel Care Long
Handschoenen
Ultra comfortable glove with long cuff and easy donning thanks to its internal polymer coating.
  • Accelerator free to limit type IV allergies
  • Great elasticity and good dexterity
  • Texture: Textured fingertips
  • Non-slip texture for excellent grip and object manipulation
  • Its robustness gives it greater durability
  • Long cuff for optimised protection
  • Made in France
Recommended in medical, industrial, and food environments.