M-Aprons, 0,5 mm
Leverancier: Basan
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
090-123 090-125 090-128
BSNC090-123EA 40.5 EUR
BSNC090-123 BSNC090-125 BSNC090-128
M-Aprons, 0,5 mm
Wegwerpkleding
M-Aprons can be utilised in various fields, such as the chemical industry, healthcare, food processing, and cleaning companies.
All aprons are machine washable up to 60 °C but should not be dry cleaned. They are not suitable for use with heat or open flames. Store them at room temperature and avoid creases by rolling them up or hanging them by the neck straps.