ESD Safety Shoes, Slip-on, JALQUARTZ
Leverancier: Jallatte
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
JIJI512/35 JIJI512/36 JIJI512/37 JIJI512/38 JIJI512/39 JIJI512/40 JIJI512/41 JIJI512/42 JIJI512/43 JIJI512/44 JIJI512/45 JIJI512/46 JIJI512/47 JIJI512/48
JALCJIJI512/35EA 128 EUR
JALCJIJI512/35 JALCJIJI512/36 JALCJIJI512/37 JALCJIJI512/38 JALCJIJI512/39 JALCJIJI512/40 JALCJIJI512/41 JALCJIJI512/42 JALCJIJI512/43 JALCJIJI512/44 JALCJIJI512/45 JALCJIJI512/46 JALCJIJI512/47 JALCJIJI512/48
ESD Safety Shoes, Slip-on, JALQUARTZ
Schoenen en laarzen
Loafer safety shoe incorporating MAXI-Soft Duo anti-fatigue technology designed for work stations that require standing for long periods (Upright or sustained activity).
  • Recycled microfiber upper
  • PU/PU sole Biomass Balance de BASF®
  • ESD/Dissipative
  • Polymer toe cap type B and midsole no metallic
  • Inner sole in non-woven textile ESD
  • Heel stiffener to reinforce the heel seat
Safety shoe protecting the electronic devices against electrostatic discharge phenomena. Upper in recycled microfibre (40% recycled polyamide). White color. Upper lined in breathable 3D Surf (66% recycled polyamide) mesh with honeycomb structure to improve the peripheral ventilation of the foot, dries out rapidly. Padded ergonomic collar in compact flexible foam, lined for extra comfort and better ankle and achilles tendon protection.
BASF® PU/PU sole BMB, BioMass Balance Certified EU-REDcert² with an urban sport design, linear profile and excellent slip resistance. Extremely light and flexible support base.
Achieve 100% substitution of fossil resources with renewable, sustainable raw materials through an independently certified approach, leading to a reduction in greenhouse gas emissions - all while maintaining the same properties and quality level as traditional safety shoes.
Certificaten: Electrostatic dissipative shoes ESD Environmental Class II tested according to EN 61340-4-3 and EN 61340-5-1.