LABOPORT® Mini Diaphragm Vacuum Pump
Leverancier: KNF
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
LABOPORT®
KNFAN8163KT18.110VEA 863 EUR
KNFAN8163KT18.110V
LABOPORT® Mini Diaphragm Vacuum Pump
Ontgassers
Chemically resistant, oil-free diaphragm vacuum pump used for a wide range of laboratory applications. It features PPS heads, PTFE-coated diaphragms and FFPM valves.
  • Ultimate vacuum (max.): 20 mbar
  • Clean, 100% oil-free operation
  • Corrosion-resistant
  • Compact, quiet and reliable
  • Small footprint
  • Maintenance-free
Application: Filtration, degassing, desiccation, fluid aspiration, solid-phase extraction.