Anti-IgG Goat Polyclonal Antibody *Cross Adsorbed* (XFD555)

Leverancier: AAT BIOQUEST
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

50162 50163
AATB50162EA 153 EUR
AATB50162 AATB50163
Anti-IgG Goat Polyclonal Antibody *Cross Adsorbed* (XFD555)
Antilichamen
Anti-human secondary antibodies have well-characterised specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target.

  • Affinity purified
  • Easily reconstituted

Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence).

Opgelet: For research use only.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: XFD555
Epitope:
Host: Goat
Isotype: IgG (H, L)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR