Storage and Assay Microplates
Leverancier: AGILENT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
203426-100 201240-100 204600-100 204357-100
AGLS203426-100EA 141 EUR
AGLS203426-100 AGLS201240-100 AGLS204600-100 AGLS204357-100
Storage and Assay Microplates
Microplaten Deepwell-microplaten
For biological assays, low storage temperatures, and filtrate collection applications.
  • Compatible with all major automation platforms and benchtop devices
  • No risk feasibility testing – test a sample in your application
  • Produced to ANSI/SLAS standards, for compatibility with popular labware and devices
  • Molded with chemical resistant polymers, for maintained performance with prolonged exposure to aggressive solutions and solvents
  • Sealable with heat and adhesive seals, or cap mats, for superior performance
  • Low temperature tolerant for refrigerator or freezer storage
  • Autoclavable, can be sterilized at 121 °C, 1,03 bar for 20 minutes
  • Available in a large range of well volumes, from 58 to 10 ml for application scale-up or scale-down
  • Complementary fit to Agilent filter microplates for centrifugation stacking
  • Manufactured in a DNAse/RNase-free ISO 9001 regulated facility
Agilent supplies collection and storage assay microplates for a wide range of reaction chemistries, biological assays, low storage temperatures, and filtrate collection applications. These microplates are molded with medical-grade, chemical-resistant polymers and are available in 24, 48, 96, or 384-well configurations of variable volumes. Agilent also supplies mat seals and lids for these microplates.