3'-O-Azidomethyl-dUTP
Leverancier: AAT BIOQUEST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
17228 17229
AATB17228EA 946 EUR
AATB17228 AATB17229
3'-O-Azidomethyl-dUTP
Reagentia voor nucleïnezuren Nucleotiden
3'-O-Azidomethyl-dUTP is an excellent dNTP building block that can be used for developing DNA detecting probes for DNA sequencing by synthesis (SBS) and other molecular biology applications.
  • Highly purified
DNA sequencing is a fundamental tool in biological and medical research (especially for personalised medicine).
Opgelet: For research use only.