Cyanine-3- dUTP [Cy3-dUTP] *1 mM in TE Buffer (pH 7,5)*
Leverancier: AAT BIOQUEST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
17025-100NM 17025-1UM
AATB17025-100NMEA 458 EUR
AATB17025-100NM AATB17025-1UM
Cyanine-3- dUTP [Cy3-dUTP] *1 mM in TE Buffer (pH 7,5)*
Reagentia voor nucleïnezuren Nucleotiden
This Cy3-dUTP conjugate can be used as a red fluorescence colour with Spectrum Orange™ filter set (Spectrum Orange™ is the trademark of Vysis).
  • Functionally equivalent to Cyanine 3-dUTP (Perkin Elmer’s NEL578001EA)
The dye-modified deoxyuridine 5'-triphosphates are one of the most common methods to produce dye-labeled DNA via conventional enzymatic incorporation methods such as reverse transcription, nick translation, random primed labelling, or PCR.
Opgelet: For research use only.