Anti-Kappa Light Chain Rat Monoclonal Antibody [clone: KT128]

Leverancier: ANTIBODIES.COM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

A164-100 A164-200
ANTIA164-100EA 445 EUR
ANTIA164-100 ANTIA164-200
Anti-Kappa Light Chain Rat Monoclonal Antibody [clone: KT128]
Antilichamen
Rat monoclonal (KT128) antibody to Ig kappa light chain for ELISA and WB with samples derived from mouse.

  • Validated applications: ELISA, WB
  • Recommended dilutions: ELISA: 1/1000, WB: 1/500 to 1/10000

Type: Primary
Antigen: Kappa Light Chain
Clonality: Monoclonal
Clone: KT128
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR