PlateLoc Thermal Microplate Sealer
Leverancier: Agilent Technologies
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AGLSG5585HAEA 40388 EUR
AGLSG5585HA
PlateLoc Thermal Microplate Sealer
Warmtesealers Warmtesealers voor microplaten
Offers reliable automated sealing for both standalone and integrated use.
  • Intuitive touch screen interface and fast cycle times with no cool-down period enable fast plate sealing
  • Automatically adjusts to accommodate a wide range of microplates and includes four types of plate stage inserts to provide support for even the most challenging microplate types
  • Energy efficient sleep mode shuts off the screen and hotplate after a designated time of inactivity to save energy
  • Quick configurability with up to 10 programmable presets to save time when changing seal settings for different sealing applications
  • Small instrument footprint and numerous proven integrations make the PlateLoc Sealer ideal for system integration
The Agilent PlateLoc Thermal Microplate Sealer is the premier fully automatic thermal heat sealer for the laboratory. Featuring an intuitive and easy-to-use interface, it accommodates deep-well, assay, PCR and compound storage microplates with a variety of seals for different applications.
This fully automated laboratory plate sealer is built on a well-established platform with a robust and compact design that is distinguished by a long and proven reputation. The PlateLoc automated microplate sealer is also ideal for robotic integration, featuring an extended-travel plate stage, RS-232 serial port, and ActiveX control.