Anti-IgG Goat Polyclonal Antibody (mFluor™ Violet 510)

Leverancier: AAT BIOQUEST
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

49050 49051
AATB49050EA 198 EUR
AATB49050 AATB49051
Anti-IgG Goat Polyclonal Antibody (mFluor™ Violet 510)
Antilichamen
mFluor™ Violet 510 goat anti-mouse conjugates are secondary antibodies designed for optimal performance in immunoassay applications, including flow cytometry, immunofluorescence, and confocal microscopy.

  • Purified and quality-tested for consistent results
  • Suitable for multiple applications
  • Purified through affinity chromatography

mFluor™ Violet 510 is optimally excited by the violet laser and emits bright green fluorescence maximally at 505 nm.

Opgelet: Research use only.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: mFluor™ Violet 510
Epitope:
Host: Goat
Isotype: IgG (H, L)
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR