ReadiUse™ ddNTP Terminator Mix *10 mM*
Leverancier: AAT BIOQUEST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
17205 17206
AATB17205EA 283 EUR
AATB17205 AATB17206
ReadiUse™ ddNTP Terminator Mix *10 mM*
Reagentia voor nucleïnezuren Nucleotiden
ReadiUse™ ddNTP Terminator Mix consists of 4 equimolar 2',3'-ddNTPs, including ddATP, ddGTP, ddTTP and ddCTP.
  • All the 2',3'-ddNTPs are highly purified
They are provided in 10 mM solution and can be readily added to a sequencing solution or other biological samples.
Opgelet: For research use only.