3'-O-Azidomethyl-dCTP
Leverancier: AAT BIOQUEST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
17222 17223
AATB17222EA 673 EUR
AATB17222 AATB17223
3'-O-Azidomethyl-dCTP
Reagentia voor nucleïnezuren Nucleotiden
3'-O-Azidomethyl-dCTP is one of the four essential dNTPs (A, C, G, T) that can be used for the DNA sequencing by synthesis (SBS).
  • Highly purified
DNA sequencing is a fundamental tool in biological and medical research (especially for personalised medicine).
Opgelet: For research use only.