Cyanine-5- dUTP [Cy5-dUTP] *1 mM in TE Buffer (pH 7,5)*
Leverancier: AAT BIOQUEST
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
17026-100NM
17026-1UM
AATB17026-100NMEA
458
EUR
AATB17026-100NM
AATB17026-1UM
Cyanine-5- dUTP [Cy5-dUTP] *1 mM in TE Buffer (pH 7,5)*
Reagentia voor nucleïnezuren
Nucleotiden
This Cy5-dUTP conjugate can be used as a red fluorescence colour with SpectrumFRed™ filter set (SpectrumFRed™ is the trademark of Vysis).
- Functionally equivalent to Cyanine 5-dUTP (Perkin Elmer’s NEL579001EA)
The dye-modified deoxyuridine 5'-triphosphates are one of the most common methods to produce dye-labelled DNA via conventional enzymatic incorporation methods such as reverse transcription, nick translation, random primed labelling, or PCR.
Opgelet: For research use only.