CytoWatch™ Wash-free Fluorescence Cell Imaging Buffer, 10X
Leverancier: AAT BIOQUEST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
20046 20045
AATB20046EA 410 EUR
AATB20046 AATB20045
CytoWatch™ Wash-free Fluorescence Cell Imaging Buffer, 10X
Koppelings- en labelingssystemen voor eiwitten
CytoWatch™ Wash-free fluorescence cell imaging buffer *10X* is a ready-to-use buffer optimised for fluorescence cell imaging.
  • Designed for ease of use
  • Saves time
CytoWatch™ Wash-free fluorescence cell imaging buffer is 10X concentrated and should be diluted to 1X with PBS before use.
Opgelet: For research use only.