CytoWatch™ Multiplexing Buffer for Flow Cytometry, 20X
Leverancier: AAT BIOQUEST
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
37010
37011
AATB37010EA
111
EUR
AATB37010
AATB37011
CytoWatch™ Multiplexing Buffer for Flow Cytometry, 20X
Buffers
Buffers voor weefselkweek
CytoWatch™ Multiplexing Buffer is a buffer optimised for the multicolor flow cytometric analysis of cells.
- Designed for ease of use
- Enhances multicolor flow experiments with two or more staining reagents
CytoWatch™ Multiplexing Buffer is a concentrated solution that is directly added to the staining solutions of certain fluorescent antibody conjugates or other fluorescent reagents before staining cells.
Opgelet: For research use only.