ANTHONY S1 Safety Shoes, Lace-Up, Low, ESD, Type 2, Elten
Leverancier: ELTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1727502/39 7275502/41 1727502/36 7275502/48 7275502/42 7275502/40 7275502/47 1727502/38 1727502/37 7275502/43 7275502/46 7275502/44 7275502/45
ELTN1727502/39EA 85.9 EUR
ELTN1727502/39 ELTN7275502/41 ELTN1727502/36 ELTN7275502/48 ELTN7275502/42 ELTN7275502/40 ELTN7275502/47 ELTN1727502/38 ELTN1727502/37 ELTN7275502/43 ELTN7275502/46 ELTN7275502/44 ELTN7275502/45
ANTHONY S1 Safety Shoes, Lace-Up, Low, ESD, Type 2, Elten
Schoenen en laarzen
Lace-up safety shoes made from a microfibre/mesh material blend.
  • Microfibre/mesh material
  • Fabric lining for natural ventilation
  • Full, padded bellows tongue
  • Semi-orthopaedic inlay black ESD sole
  • Soft fleece ESD insole
  • TPU/PU sole ERGO-ACTIVE 2
  • Steel toe cap
Designed for use in dry work areas, such as industry, storage, transport, and assembly (S1 environments). These safety shoes are also suitable for use in areas where there is a risk of electrostatic discharge (ESDS/ESD).
Certificaten: EN ISO 20345:2022 S1 FO, form A.