Anti-Tartrate Resistant Acid Phosphatase Mouse Monoclonal Antibody [clone: ABT-TRAP]

Leverancier: ANTIBODIES.COM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

A100013-100 A100013-50
ANTIA100013-100EA 475 EUR
ANTIA100013-100 ANTIA100013-50
Anti-Tartrate Resistant Acid Phosphatase Mouse Monoclonal Antibody [clone: ABT-TRAP]
Antilichamen
Mouse monoclonal (ABT-TRAP) antibody to TRAP for IHC with samples derived from human.

  • Validated Applications: IHC

Type: Primary
Antigen: Tartrate Resistant Acid Phosphatase
Clonality: Monoclonal
Clone: ABT-TRAP
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR