6-VIC phosphoramidite
Leverancier: AAT BIOQUEST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
6086 6087 6088
AATB6086EA 341 EUR
AATB6086 AATB6087 AATB6088
6-VIC phosphoramidite
6-VIC phosphoramidite
6-VIC phosphoramidite is an asymmetrical xanthene dye widely used to prepare fluorescence-labeled oligonucleotides and design qPCR probes, such as TaqMan, Molecular Beacon, and Scorpion.
  • Stable for 6 months
Quality tested in-house
Opgelet: For Research Use Only
New Product
Order Now
Specificaties
Environmentally Preferable