Fluorescein-dT CPG
Leverancier: AAT BIOQUEST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
6205 6206
AATB6205EA 138 EUR
AATB6205 AATB6206
Fluorescein-dT CPG
Fluorescein-dT CPG
Fluorescein CPG has traditionally been used to add the fluorescein label at the 3'-terminus.
  • Stable for 12 months
Quality tested in-house