iFluor® Ultra 594 succinimidyl ester
Leverancier: AAT BIOQUEST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
71650 71651 71652
AATB71650EA 429 EUR
AATB71650 AATB71651 AATB71652
iFluor® Ultra 594 succinimidyl ester
iFluor® Ultra 594 succinimidyl ester
iFluor® Ultra 594 is an exceptionally bright red fluorescent dye excitable by 561 nm or 594 nm laser lines, with a peak emission at 601 nm.
  • Stable for 12 months
Fluorescent dye-conjugated antibodies provide a tool for identifying proteins in many applications including fluorescent cell imaging, flow cytometry, western blotting, immunohistochemistry and more.
Opgelet: For Research Use Only