APC maleimide, ReadiUse™, preactivated [Activated Allophycocyanin]
Leverancier: AAT BIOQUEST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2567 2568
AATB2567EA 206 EUR
AATB2567 AATB2568
APC maleimide, ReadiUse™, preactivated [Activated Allophycocyanin]
APC maleimide
ReadiUse™ Preactivated APC Maleimide is prepared by reacting highly purified allophycocyanin (APC) with SMCC.
  • Stable for 6 months
Quality tested in-house
Opgelet: For Research Use Only