PE maleimid, ReadiUse™, preactivated [Activated R-Phycoerythrin]
Leverancier: AAT BIOQUEST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2565 2566
AATB2565EA 136 EUR
AATB2565 AATB2566
PE maleimid, ReadiUse™, preactivated [Activated R-Phycoerythrin]
PE maleimid
ReadiUse™ Preactivated PE Maleimide is prepared by reacting highly purified R-Phycoerythrin (PE) with SMCC.
  • Stable for 6 months
Quality tested in-house
Opgelet: For Research Use Only