SoNa™ 520 AM
Leverancier: AAT BIOQUEST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
21323 21324
AATB21323EA 687 EUR
AATB21323 AATB21324
SoNa™ 520 AM
SoNa™ 520 AM
SoNa™ 520 AM is the cell-permeable version of SoNa™ 520 used for monitoring sodium ions in live cells.
  • Stable for 12 months
Quality tested in-house
Opgelet: For Research Use Only