Anti-IgG Goat Polyclonal Antibody (iFluor® 800)
Leverancier: AAT BIOQUEST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
48000 48001
AATB48000EA 138 EUR
AATB48000 AATB48001
Anti-IgG Goat Polyclonal Antibody (iFluor® 800)
Antilichamen
AAT Bioquest's iFluor® dyes are optimised for labeling proteins, in particular, antibodies.
  • Purified and quality-tested for consistent results
  • Suitable for multiple applications
  • Purified through affinity chromatography
iFluor® 800 goat anti-mouse IgG (H+L) conjugate has fluorescence excitation and emission maxima of ∼801 nm and ∼820 nm, respectively.
Opgelet: Research use only.
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: iFluor® 800
Epitope:
Host: Goat
Isotype: IgG (H, L)
Reactivity: Mouse