Chemical-Resistant PP Labels B-7425J for BradyJet™ J4000
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
152783 152784 152785 152786 152778 152779 152780 152781 152782
BRDY152783EA 256.24 EUR
BRDY152783 BRDY152784 BRDY152785 BRDY152786 BRDY152778 BRDY152779 BRDY152780 BRDY152781 BRDY152782
Chemical-Resistant PP Labels B-7425J for BradyJet™ J4000
Etiketten
This B-7425J polypropylene material is ideal for GHS and other full-colour identification applications where resistance to harsh chemicals is required.
  • Matt white polypropylene with acrylic adhesive is inkjet printable
  • Low-cost, full-colour GHS labeling option withstands chemical exposure
  • Service temperature of -80 to 70°C
  • An ink cartridge is required to print this material; use Part Number J40-CMY
For component and equipment labelling, general identification, hazard communication identification, laboratory labelling.