ReadiPrep™ Nickel NTA Magnetic Beads
Leverancier: AAT BIOQUEST
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
56000
56001
AATB56000EA
130
EUR
AATB56000
AATB56001
ReadiPrep™ Nickel NTA Magnetic Beads
Eiwitzuivering
Reagentia voor eiwitpurificatie
ReadiPrep™ Nickel NTA magnetic beads provide a fast and simple solution for small-scale affinity purification of histidine-tagged proteins.
- Stable for 12 months
Quality tested in-house.
Opgelet: For research use only.