Beadlite™ Rapid Colorimetric Maleimide Quantitation Kit for Nanoparticles
Leverancier: AAT BIOQUEST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5534 5533
AATB5534EA 402 EUR
AATB5534 AATB5533
Beadlite™ Rapid Colorimetric Maleimide Quantitation Kit for Nanoparticles
Testen Enzyme-assays
Designed for consistent and dependable results. The Beadlite™ Rapid Colorimetric Maleimide Quantitation Kit is an essential tool for researchers involved in bioconjugation research, specifically designed for the accurate quantification of maleimide functional groups on solid surfaces such as magnetic beads, polystyrene beads, or other microsphere nanoparticles.
  • Each kit provides sufficient reagents for 100 assays, with a volume of 0,5 ml per assay.
  • Consistent results
  • Six-month shelf life
Maleimide-thiol chemistry is integral to the engineering of antibody-nanoparticle (NP) conjugates, with nanoparticle surface activation typically achieved via SMCC-mediated maleimide functionalization.
Leveringsinformatie: Shipped with blue ice.
Opgelet: Research use only.