ReadiPrep™ Protein G-Agarose Resin
Leverancier: AAT BIOQUEST
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
55016
55017
AATB55016EA
133
EUR
AATB55016
AATB55017
ReadiPrep™ Protein G-Agarose Resin
Eiwitzuivering
Protein Purification Resins
ReadiPrep™ Protein G-Agarose Resin is a high-quality affinity resin designed for the isolation and purification of immunoglobulin classes, subclasses, and fragments from mammalian serum and biological fluids.
- Stable for 12 months
Quality tested in-house.
Opgelet: For research use only.