CRYO-LNG™ Gloves
Leverancier: TEMPSHIELD
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CLMLWP CLMMWP CLMXWP CLMXXWP
TEMPCLMLWPEA 412 EUR
TEMPCLMLWP TEMPCLMMWP TEMPCLMXWP TEMPCLMXXWP
CRYO-LNG™ Gloves
Handschoenen
Hand protection for industrial settings working with LNG or liquid nitrogen.
  • Abrasion resistant palm fabric safeguards against rough surfaces
  • A thin, seamless 100% waterproof liner provides protection from spills and splashes
  • Cryogenic protection for ultra-cold applications down to –196 °C (–320 °F)
  • High performance, thermal inner lining wicks moisture away from hands, maintaining comfort over extended periods
Gloves are designed to provide a high level of thermal and splash protection where the exposure to cryogenic fluids might exist.
State-of-the art materials and a multi-layered construction allow for a maximum level of thermal protection, flexibility, and dexterity which are essential features when function is important and safety is critical.
Applications: Delivering cryogenic gases, servicing cryogenic systems, refueling and servicing at LNG filling stations, working in LNG transportation and facilities, working in mining, oil, and gas.
Certificaten: EN 511, EN 388, EN ISO 4120
Opgelet: Not intended for immersion in liquid nitrogen or other cryogenic liquids.