Spectrophotometer Cuvettes, TrayCell 2.0
Leverancier: HELLMA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
105830-A3-V1-4 105830-A1-V1-4
HELL105830-A3-V1-4EA 0 EUR
HELL105830-A3-V1-4 HELL105830-A1-V1-4
Spectrophotometer Cuvettes, TrayCell 2.0
Cuvetten Cuvetten voor spectrofotometers
Suitable for all current spectrophotometers with standard cuvette holder.
  • Samples with low surface tension can be measured
  • Excellent reproducibility
  • Quick and easy to clean
  • Strictly defined, precise path lengths: misadjustment impossible
  • Path lengths in delivery included: 1,0 and/or 0,2 mm
  • Additional path lengths: 2,0 and 0,1 mm (optional available)
W×D: 12,5×12,5 mm
Max. temperature: 50 °C
Fiber optics: Built in not exchangeable, low solarisation 210 to 1100 nm
Large measuring range from approximately 6 to 8500 ng/μl (dsDNA) and approximately 0,1 toMore Product Information