Eppendorf® Multi-Channel Pipette Starter Sets with 50 Racks of ep Dualfilter T.I.P.S.® 384 Filter Tips
Leverancier: EPPENDORF
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3125000.915 3125000.923 3125000.931 3125000.940 4861000.952 4861000.953 4861000.954 4861000.955
613-7317EA 3370 EUR
613-7317 613-7318 613-7319 613-7320 613-7321 613-7322 613-7323 613-7324
Eppendorf® Multi-Channel Pipette Starter Sets with 50 Racks of ep Dualfilter T.I.P.S.® 384 Filter Tips
Pipetten Micropipetten
Transition from the tedious process of alternate well pipetting or vessel reformatting with single-channel pipettes to a more efficient manual liquid handling with 384-well plates with one of these starter sets. Each starter set includes a mechanical Eppendorf Research® Plus or electronic Eppendorf Xplorer® Plus multi-channel pipette and a total of 19200 ep Dualfilter T.I.P.S.® 384 filter tips (50 racks of 384 tips) in 'PCR clean and Sterile' grade.
  • Available for a limited time only
  • Starter sets offered for Eppendorf Research® Plus or Eppendorf Xplorer®