VWR® Sterile Nitrile Cleanroom Class 10 Gloves
Leverancier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
112-6211EA 455 EUR
112-6211 112-6212 112-6213 112-6214 112-6215 112-6216 112-6217
VWR® Sterile Nitrile Cleanroom Class 10 Gloves
Handschoenen
Powder-free gloves designed to provide comfort and tactile sensitivity.
  • Hand-specific, beaded cuff
  • Silicone-free
  • Low particle count
  • Specialized manufacturing process
VWR® Sterile nitrile cleanroom class 10 gloves are entirely latex-free to prevent the risk of allergic reactions. These gloves are supplied sterile.
Certificaten: EN ISO 21420:2020, EN ISO 374-1:2016 Type B (KPT), EN ISO 374-5:2016; CAT III; AQL 1.5
Verpakking: Each pair is double packaged in a wallet within a pouch, 20 pouches per bag, 10 bags per case (200 pairs).