Hollow Stoppers [ST]
Leverancier: witeg Labortechnik
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2.436.000 2.436.001
WITG2.436.000EA 25.7 EUR
WITG2.436.000 WITG2.436.001
Hollow Stoppers [ST]
Stoppen Glazen stoppen
ST-hollow stoppers, with extra long stem, for BOD-bottles, made of borosilicate glass.