VWR® Rigid qPCR Plates, 96-Well, with White Wells
Leverancier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
732-4403EA 66.1 EUR
732-4403 732-4405
VWR® Rigid qPCR Plates, 96-Well, with White Wells
Microplaten PCR-platen
Rigid qPCR Plates feature a rugged polycarbonate frame, providing a solid and flat surface that guarantees seamless interaction with automated liquid handling platforms. With a firm grip assured, robots can confidently place these plates with utmost precision, eliminating any concerns of misalignment.
  • Polycarbonate frame is rigid and flat, facilitating a firm grip by robotic arms and ensuring reliable placement
  • Polypropylene wells provide optimal thermal transfer and higher reaction efficiency
  • White wells enhance fluorescence signals during qPCR
  • Wells are slightly raised to ensure evaporation-proof sealing with film, foil, or strip caps
  • ANSI/SBS footprint and well spacing
  • Vibrant colors offer a visual distinction between different samples or experiments
VWR® Rigid qPCR Plates feature no-flex polycarbonate frames and polypropylene wells designed to facilitate optimal thermal transfer and higher reaction efficiency. This ensures more accurate and reliable results, providing researchers with the confidence they need in their experiments. Additionally, white well options are carefully designed to enhance fluorescence signals during qPCR, enabling clearer and more precise measurements.
Sealing qPCR plates is a critical aspect of laboratory workflows, and VWR® excels in this area. The slightly raised wells of VWR® Rigid qPCR Plates ensure an evaporation-proof seal when used with film, foil, or strip caps. Say goodbye to concerns about sample integrity or loss due to evaporation. VWR® Rigid qPCR Plates keep valuable samples protected throughout the entire process.
With an ANSI/SBS footprint and well spacing, VWR® Rigid qPCR Plates integrate seamlessly into existing laboratory setups, offering compatibility and ease of use. Plus, the vibrant color options allow for quick and efficient identification and organization by providing visual distinctions between different samples or experiments.
Certificaten:
Free of DNase, RNase, DNA, and PCR inhibitors.
Verpakking:
10 plates per pack, 10 packs per case.