perform® classic Surface Disinfectant, Concentrate OXY
Leverancier: Schülke
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
perform® classic
SCUE70003330EA 0 EUR
SCUE70003330
perform® classic Surface Disinfectant, Concentrate OXY
Reinigingsmiddelen Desinfectiemiddelen
Powdered concentrate based on active oxygen for disinfecting surfaces.
  • Sporicidal and material compatible
  • Convenient single-use sachets
  • Colour free
  • Granular form
Disinfection of all types of surfaces. perform® classic concentrate OXY is suitable for use in all high-risk areas where, in addition to broad effectiveness, strict requirements are placed on the environmental behaviour and user-friendliness of a surface disinfectant. Due to the excellent material compatibility of the active solution, perform® classic concentrate OXY can also be used with problematic materials, such as acrylic glass (incubators).
Composition:
100 g powder contains the following active substances: 45 g Pentapotassium bis (peroximonosulphate) bis (sulphate)
Labelling according to regulation (EC) No. 648/2004: 5 to 15% anionic surfactants, <5% non-ionic surfactants, <5% soap, <5% phosphonates, perfumes
Certificaten: Efficacy tested according to European norms (EN).
Opgelet: Use disinfectants safely. Always read the label and product information before use. Do not store bags above room temperature. Wearing of protective gloves (e.g. made of Nitrile rubber) is recommended. The active solution may bleach textile dyes that are highly sensitive.