NucSpot® nuclear stain
Leverancier: Biotium
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
41038 41038-T
41038.EA 668 EUR
41038. 41038-T
NucSpot® nuclear stain
NucSpot®
Cell membrane-impermeant, nuclear-specific counterstains. Suitable for fixed cells or staining dead cells in live cultures.