Circular Vinyl Labels for BradyJet™ J7300 Inkjet Label Printer
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
170476 170477
BRDY170476EA 103.66 EUR
BRDY170476 BRDY170477
Circular Vinyl Labels for BradyJet™ J7300 Inkjet Label Printer
Etiketten
General-purpose vinyl labels feature a specially formulated topcoat for improved durability and colour. For use on smooth and textured and rough surfaces.
  • Tested to withstand exposure to liquids, alcohol and chemicals, as well as indoor fluorescent and UV lighting
  • Works well in low temperatures with a service temperature of −40 to 60 °C
  • Inkjet receptive vinyl material
  • Designed for indoor/outdoor use
  • Abrasion-resistant, oil-resistant, outdoor durability, solvent/chemical resistant
  • Matt finish
Low-shrink vinyl with ultra-aggressive adhesive is ideal forMore Product Information