Brady® UL Approved Polyester Label Tape for BradyJet™ J7300 Colour Label Printer
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
176574 176576 176575
BRDY176574EA 229.55 EUR
BRDY176574 BRDY176576 BRDY176575
Brady® UL Approved Polyester Label Tape for BradyJet™ J7300 Colour Label Printer
Etiketten
Polyester material meets UL 969 standard for marking and labelling and is ideal for product labelling, logos, 'Right to Know' labels, and barcode applications.
  • Durable and resistant to abrasions, chemicals, oils, and solvents
  • Strong acrylic adhesive works best on flat, smooth surfaces
  • Inkjet receptive polyester material
  • Designed for indoor use
Inkjet-receptive labels allows label creation with variable data and colour components in-house. Built-in LabelSense™ technology enables cartridge and printer communication for fast changeovers, automatic labelMore Product Information