Cryogenic Polyester Labels FreezerBondz™ B-2492 for BradyJet™ J7300 Inkjet Printer
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
176646 176642 176647 176644 176643 176641 176645
BRDY176646EA 476.39 EUR
BRDY176646 BRDY176642 BRDY176647 BRDY176644 BRDY176643 BRDY176641 BRDY176645
Cryogenic Polyester Labels FreezerBondz™ B-2492 for BradyJet™ J7300 Inkjet Printer
Etiketten
Trusted FreezerBondz™ material Polyester B-2492 combines high performance with the power of Brady inkjet printing for lab work involving curved and frozen surfaces. For use on smooth surfaces.
  • Specially designed topcoat protects vibrant colours from harsh chemicals
  • Withstands chemicals and extreme temperatures, including storage in liquid nitrogen
  • Top-performing adhesive allows for tight bonding onto smooth, curved surfaces
  • Can be applied to frozen surfaces and protects sample contents from unnecessary thaw cycles
  • Designed for indoor use
  • Alcohol-resistant, low temperature resistant, solvent/chemical resistant
  • Matt finish
Full-colour printing allows for faster identification of individual or group samples, and makes lab techs aware of important information during sample processing. Top-performing adhesive allows for tight bonding onto smooth, curved surfaces including test tubes, vials, and straw pipettes. Built-in LabelSense™ technology enables cartridge and printer communication for fast changeovers, automatic label setup, and seamless software interaction.
For use with the BradyJet™ J7300 Colour Label Printer. Ink cartridges are required to print on this material; use part numbers P-CMY-1 and P-K-1.