Brady® Repositionable Vinyl Labels for BradyJet™ J7300 Colour Label Printer
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
170456 170455 170457
BRDY170456EA 336.93 EUR
BRDY170456 BRDY170455 BRDY170457
Brady® Repositionable Vinyl Labels for BradyJet™ J7300 Colour Label Printer
Etiketten
Specialised aggressive adhesive allows sticking, removing, and repositioning labels with ease without leaving behind adhesive residue or surface damage.
  • Durable, flexible material is purpose-built for areas where identification needs changed frequently
  • Works well in low temperatures with a service temperature of −40 to +60 °C
  • Repositionable inkjet receptive vinyl material
  • Designed for indoor use
Durable, flexible inkjet receptive vinyl material is ideal for Lean/5S and work in process labelling as well as warehousing racks, bins, and storage. Built-in LabelSense™ technology enables cartridge and printer communication for fast changeovers, automatic label setup, and seamless software interaction.
For use with the BradyJet™ J7300 Colour Label Printer. Ink cartridges are required to print on this material; use part numbers P-CMY-1 and P-K-1.