Brady® Vinyl Label Tapes for BradyJet™ J7300 Colour Label Printer
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
176580 176582 170458 176581
BRDY176580EA 231.4 EUR
BRDY176580 BRDY176582 BRDY170458 BRDY176581
Brady® Vinyl Label Tapes for BradyJet™ J7300 Colour Label Printer
Etiketten
General-purpose vinyl labels feature a specially formulated topcoat for improved durability and colour.
  • Low-shrink vinyl with ultra-aggressive adhesive is versatile for a wide array of applications
  • Tested to withstand exposure to liquids, alcohol and chemicals, as well as indoor fluorescent and UV lighting
  • Works well in low temperatures with a service temperature of −40 to +60 °C
  • Inkjet receptive vinyl material
  • Designed for indoor/outdoor use
General-purpose vinyl labels are durable up to two years outdoors, even longer indoors. Ideal for Lean, general facility, pipe marking, OSHA/ANSI, PPE, machine labels and tags, quality and control, lockout, GHS and chemical, warehousing, general laboratory and more. Enhanced topcoat works with durable J7300 inks to protect printed text and graphics from smudging and harsh conditions. Built-in LabelSense™ technology enables cartridge and printer communication for fast changeovers, automatic label setup and seamless software interaction.
For use with the BradyJet™ J7300 Colour Label Printer. Ink cartridges are required to print on this material; use part numbers P-CMY-1 and P-K-1.