Brady® Water Soluble Paper Label Tapes for BradyJet™ J7300 Colour Label Printer
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
170461 176583 170460 176605 170462
BRDY170461EA 99.96 EUR
BRDY170461 BRDY176583 BRDY170460 BRDY176605 BRDY170462
Brady® Water Soluble Paper Label Tapes for BradyJet™ J7300 Colour Label Printer
Etiketten
Choose water soluble paper labels for complete label and adhesive removal. Ideal for laboratories, temporary identification needs, and work-in-process product flows.
  • Label and adhesive completely remove in 30 seconds using warm water
  • Inkjet receptive labels resist abrasions, humidity, and temperatures from −70 to +100 °C
  • Designed for indoor use
Dissolvable acrylic adhesive is ideal for labs, asset tracking, and temporary ID. Built-in LabelSense™ technology enables cartridge and printer communication for fast changeovers, automatic label setup, and seamless software interaction.
For use with the BradyJet™ J7300 Colour Label Printer. Ink cartridges are required to print on this material; use part numbers P-CMY-1 and P-K-1.
Certificaten: RoHS compliant to 2011/65/EU (Pass 2015/863).